missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Tryptophan, Cell Culture Reagent

CAS: 73-22-3 | C11H12N2O2 | 204.23 g/mol

$59.49 - $1306.33

Chemical Identifiers

CAS 73-22-3
Molecular Formula C11H12N2O2
Molecular Weight (g/mol) 204.23
MDL Number MFCD00064340
InChI Key QIVBCDIJIAJPQS-VIFPVBQESA-N
Synonym l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin
PubChem CID 6305
ChEBI CHEBI:16828
IUPAC Name (2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
SMILES N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6250809
View Documents
Thermo Scientific Chemicals
J6250809
10 g
Each for $59.49
Only null left
 
AAJ6250822
View Documents
Thermo Scientific Chemicals
J6250822
100 g
Each for $252.44
Only null left
 
AAJ62508A1
View Documents
Thermo Scientific Chemicals
J62508A1
1 kg
Each for $1,306.33
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

73-22-3
204.23
QIVBCDIJIAJPQS-VIFPVBQESA-N
6305
(2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
C11H12N2O2
MFCD00064340
l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin
CHEBI:16828
N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

280°C to 285°C
White
MFCD00064340
14,9797
QIVBCDIJIAJPQS-VIFPVBQESA-N
(2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
204.23
CHEBI:16828
Cell Culture Reagent
L-Tryptophan
73-22-3
C11H12N2O2
86197
l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin
N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
10 g
6305
204.23
Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-795-6

RTECSNumber : YN6130000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only