missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-(+)-Mandelic Acid 99.0+%, TCI America™
$132.35 - $656.14
Chemical Identifiers
CAS | 17199-29-0 |
---|---|
Molecular Formula | C8H8O3 |
Molecular Weight (g/mol) | 152.149 |
MDL Number | MFCD00004495 |
InChI Key | IWYDHOAUDWTVEP-ZETCQYMHSA-N |
Synonym | s-+-mandelic acid, s-mandelic acid, s-2-hydroxy-2-phenylacetic acid, l-mandelic acid, l-+-mandelic acid, 2s-2-hydroxy-2-phenylacetic acid, mandelic acid, s, unii-l0umw58g3t, l +-mandelic acid |
PubChem CID | 439616 |
ChEBI | CHEBI:32800 |
IUPAC Name | (2S)-2-hydroxy-2-phenylacetic acid |
SMILES | C1=CC=C(C=C1)C(C(=O)O)O |
Chemical Identifiers
17199-29-0 | |
152.149 | |
IWYDHOAUDWTVEP-ZETCQYMHSA-N | |
439616 | |
(2S)-2-hydroxy-2-phenylacetic acid |
C8H8O3 | |
MFCD00004495 | |
s-+-mandelic acid, s-mandelic acid, s-2-hydroxy-2-phenylacetic acid, l-mandelic acid, l-+-mandelic acid, 2s-2-hydroxy-2-phenylacetic acid, mandelic acid, s, unii-l0umw58g3t, l +-mandelic acid | |
CHEBI:32800 | |
C1=CC=C(C=C1)C(C(=O)O)O |
Specifications
2.3 (10 g/L aq. soln.) | |
17199-29-0 | |
C8H8O3 | |
s-+-mandelic acid, s-mandelic acid, s-2-hydroxy-2-phenylacetic acid, l-mandelic acid, l-+-mandelic acid, 2s-2-hydroxy-2-phenylacetic acid, mandelic acid, s, unii-l0umw58g3t, l +-mandelic acid | |
C1=CC=C(C=C1)C(C(=O)O)O | |
152.149 | |
CHEBI:32800 | |
≥99.0% (GC,T) | |
Crystalline Powder |
134°C | |
White to Yellow | |
MFCD00004495 | |
IWYDHOAUDWTVEP-ZETCQYMHSA-N | |
(2S)-2-hydroxy-2-phenylacetic acid | |
439616 | |
152.15 | |
25 g | |
L-(+)-Mandelic Acid |
Safety and Handling
EINECSNumber : (3)-1764
TSCA : Yes