Learn More
Thermo Scientific Chemicals L-Lysine monohydrochloride, 99+%
CAS: 657-27-2 | C6H15ClN2O2 | 182.65 g/mol
$41.12 - $220.66
Chemical Identifiers
CAS | 657-27-2 |
---|---|
Molecular Formula | C6H15ClN2O2 |
Molecular Weight (g/mol) | 182.65 |
MDL Number | MFCD00064564 MFCD00081870 |
InChI Key | BVHLGVCQOALMSV-JEDNCBNOSA-N |
Synonym | l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl |
PubChem CID | 69568 |
ChEBI | CHEBI:53633 |
IUPAC Name | (2S)-2,6-diaminohexanoic acid;hydrochloride |
SMILES | Cl.NCCCC[C@H](N)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1624918
|
Thermo Scientific Chemicals
A1624918 |
50 g |
Each for $41.12
|
|
|||||
AAA1624930
|
Thermo Scientific Chemicals
A1624930 |
250 g |
Each for $96.56
|
|
|||||
AAA162490B
|
Thermo Scientific Chemicals
A162490B |
1000 g |
Each for $220.66
|
|
|||||
Description
L-Lysine monohydrochloride is widely used as nutritional supplements in food and beverage industries. It can also be used in animal feed as source of L-Lysine. L-Lysine Monohydrochloride can be used in a wide variety of industries including: food production, beverage, pharmaceutical, agriculture/animal feed, and various other industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Lysine monohydrochloride is widely used as nutritional supplements in food and beverage industries. It can also be used in animal feed as source of L-Lysine. L-Lysine Monohydrochloride can be used in a wide variety of industries including: food production, beverage, pharmaceutical, agriculture/animal feed, and various other industries.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Chemical Identifiers
657-27-2 | |
182.65 | |
BVHLGVCQOALMSV-JEDNCBNOSA-N | |
69568 | |
(2S)-2,6-diaminohexanoic acid;hydrochloride |
C6H15ClN2O2 | |
MFCD00064564 MFCD00081870 | |
l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl | |
CHEBI:53633 | |
Cl.NCCCC[C@H](N)C(O)=O |
Specifications
∼260°C (decomposition) | |
C6H15ClN2O2 | |
3563889 | |
l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl | |
BVHLGVCQOALMSV-JEDNCBNOSA-N | |
(2S)-2,6-diaminohexanoic acid;hydrochloride | |
182.65 | |
CHEBI:53633 | |
182.65 | |
L-Lysine monohydrochloride |
657-27-2 | |
MFCD00064564 MFCD00081870 | |
14,5636 | |
It is soluble in water. | |
Cl.NCCCC[C@H](N)C(O)=O | |
50 g | |
69568 | |
+20° (c=5 in 5N HCl) | |
≥99% |
Safety and Handling
EINECSNumber : 211-519-9
RTECSNumber : OL5650000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only