missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ L-Lysine, Hydrochloride, Calbiochem™,
$77.10 - $195.00
Chemical Identifiers
CAS | 657-27-2 |
---|---|
Molecular Formula | C6H15ClN2O2 |
Molecular Weight (g/mol) | 182.65 |
MDL Number | MFCD00064564 MFCD00081870 |
InChI Key | BVHLGVCQOALMSV-JEDNCBNOSA-N |
Synonym | l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl |
PubChem CID | 69568 |
ChEBI | CHEBI:53633 |
IUPAC Name | (2S)-2,6-diaminohexanoic acid hydrochloride |
SMILES | Cl.NCCCC[C@H](N)C(O)=O |
Chemical Identifiers
657-27-2 | |
182.65 | |
BVHLGVCQOALMSV-JEDNCBNOSA-N | |
69568 | |
(2S)-2,6-diaminohexanoic acid hydrochloride |
C6H15ClN2O2 | |
MFCD00064564 MFCD00081870 | |
l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl | |
CHEBI:53633 | |
Cl.NCCCC[C@H](N)C(O)=O |
Specifications
657-27-2 | |
C6H15ClN2O2 | |
MFCD00064564 MFCD00081870 | |
BVHLGVCQOALMSV-JEDNCBNOSA-N | |
(2S)-2,6-diaminohexanoic acid hydrochloride | |
182.65 | |
CHEBI:53633 | |
Solid |
White | |
Glass Bottle | |
l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl | |
Cl.NCCCC[C@H](N)C(O)=O | |
100 g | |
69568 | |
≥98% (Titration (dry basis)) |