missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-Isoleucine, Cell Culture Reagent
CAS: 73-32-5 | C6H13NO2 | 131.18 g/mol
$110.49 - $308.93
Chemical Identifiers
CAS | 73-32-5 |
---|---|
Molecular Formula | C6H13NO2 |
Molecular Weight (g/mol) | 131.18 |
MDL Number | MFCD00064222 MFCD00004268 |
InChI Key | AGPKZVBTJJNPAG-WHFBIAKZSA-N |
Synonym | l-isoleucine, isoleucine, 2s,3s-2-amino-3-methylpentanoic acid, s-isoleucine, s,s-isoleucine, 2s,3s-isoleucine, 2-amino-3-methylvaleric acid, erythro-l-isoleucine, l-+-isoleucine, l-ile |
PubChem CID | 6306 |
ChEBI | CHEBI:17191 |
IUPAC Name | (2S,3S)-2-amino-3-methylpentanoic acid |
SMILES | CC[C@H](C)[C@H](N)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6304514
|
Thermo Scientific Chemicals
J6304514 |
25 g |
Each for $110.49
|
|
|||||
AAJ6304522
|
Thermo Scientific Chemicals
J6304522 |
100 g |
Each for $308.93
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
73-32-5 | |
131.18 | |
AGPKZVBTJJNPAG-WHFBIAKZSA-N | |
6306 | |
(2S,3S)-2-amino-3-methylpentanoic acid |
C6H13NO2 | |
MFCD00064222 MFCD00004268 | |
l-isoleucine, isoleucine, 2s,3s-2-amino-3-methylpentanoic acid, s-isoleucine, s,s-isoleucine, 2s,3s-isoleucine, 2-amino-3-methylvaleric acid, erythro-l-isoleucine, l-+-isoleucine, l-ile | |
CHEBI:17191 | |
CC[C@H](C)[C@H](N)C(O)=O |
Specifications
∽287°C (decomposition) | |
White | |
MFCD00064222 MFCD00004268 | |
14,5179 | |
AGPKZVBTJJNPAG-WHFBIAKZSA-N | |
(2S,3S)-2-amino-3-methylpentanoic acid | |
131.18 | |
CHEBI:17191 | |
131.18 | |
Powder |
73-32-5 | |
C6H13NO2 | |
1721792 | |
l-isoleucine, isoleucine, 2s,3s-2-amino-3-methylpentanoic acid, s-isoleucine, s,s-isoleucine, 2s,3s-isoleucine, 2-amino-3-methylvaleric acid, erythro-l-isoleucine, l-+-isoleucine, l-ile | |
CC[C@H](C)[C@H](N)C(O)=O | |
25 g | |
6306 | |
+40° (c=5 in 6N HCl) | |
Cell Culture Reagent | |
L-Isoleucine |
Safety and Handling
EINECSNumber : 200-798-2
RTECSNumber : NR4705000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only