missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLRG1 Monoclonal Antibody (2F1), Brilliant Violet™ 711, eBioscience™, Invitrogen™
Syrian Hamster Monoclonal Antibody
$607.48 - $1286.23
Specifications
Antigen | KLRG1 |
---|---|
Clone | 2F1 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
40-758-9380
|
Invitrogen™
407589380 |
25 μg |
Each for $607.48
|
|
|||||
40-758-9382
|
Invitrogen™
407589382 |
100 μg |
Each for $1,286.23
|
|
|||||
Description
KLRG1 Monoclonal antibody specifically detects KLRG1 in Mouse samples. It is validated for Flow CytometrySpecifications
KLRG1 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
2F1; 2F1-Ag; CLEC15A; C-type lectin domain family 15 member A; C-type lectin domain family 15, member A; ITIM-containing receptor MAFA-L; killer cell lectin like receptor G1; killer cell lectin-like receptor subfamily G member 1; killer cell lectin-like receptor subfamily G, member 1; Klrg1; Mafa; MAFA-2F1; MAFAL; MAFA-L; MAFA-LIKE; MAFA-like receptor; Mast cell function-associated antigen; mast cell function-associated antigen (ITIM-containing); mast cell function-associated antigen 2F1; natural killer cell receptor | |
IgG | |
Affinity Chromatography | |
Antibody |
2F1 | |
Flow Cytometry | |
Brilliant Violet 711 | |
Syrian Hamster | |
Mouse | |
O88713 | |
50928 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
Klrg1 |