Learn More
DWK Life Sciences Kimble™ KIMAX™ Glass Tip
Threaded buret tip
Supplier: DWK Life Sciences 17034H99
Description
- For 03-700-12 Series and 03-857 Series burets
- Fits 25 and 50mL sizes
Specifications
17021H and 17121H Straight Bore PTFE Stopcock Burettes | |
PTFE | |
Calibrated to Accurately Deliver Buret Contents at a Proper Rate | |
Buret Tip |