Learn More
DWK Life Sciences Kimble™ Clear Glass French Squares with PTFE-Faced LDPE Foam-lined Caps
Wide mouth design for efficient addition or removal of contents
$153.15 - $298.21
Specifications
Color Clear
Material Soda Lime Glass
Closure Color Black
Product Type French Square Bottle
Closure Material Phenolic
View More Specs
Products 6
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
13-756-356
View Documents
DWK Life Sciences
5611220V26
15 mL
Case of 48 Each for $178.92
Only null left
13-756-357
View Documents
DWK Life Sciences
5610124V26
30 mL
Case of 48 Each for $298.21
Only null left
13-756-358
View Documents
DWK Life Sciences
5610228V26
60 mL
Case of 48 Each for $253.18
Only null left
13-756-359
View Documents
DWK Life Sciences
5610433V26
125 mL
Case of 24 Each for $153.15
Only null left
13-756-360
View Documents
DWK Life Sciences
5610843V26
250 mL
Case of 24 Each for $197.41
Only null left
13-756-361
View Documents
DWK Life Sciences
5611648V26
500 mL
Case of 24 Each for $293.24
Only null left
Description
Description
  • French square bottles are ideal for mixing, storing and sampling liquids or solid matter
  • Square shape maximizes use of shelf space
  • Clear glass allows for easy viewing of contents
Specifications
Specifications
Clear
Black
Phenolic
Soda Lime Glass
French Square Bottle
Screw Cap
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'