missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark Professional™ WypAll™ X60 Wipers
Fast absorption and outstanding tear resistance
Supplier: Kimberly-Clark Professional™ 35411
Description
Fast absorption and outstanding tear resistance
- Made from Hydroknit™, an advanced wiper material manufactured using jets of water to bond soft, absorbent paper to extra-strong, nonwoven fabric
- Reinforced for extra scrubbing and cleaning power
- No adhesives or binders—soft enough for face and hands
Specifications
Cleaning Cloth | |
25 cm | |
31 cm | |
Blue | |
Small Roll, Blue |
9.8 in. | |
12.2 in. | |
Non-woven Polyproylene Cellulose | |
9.8 x 12.2 in. (25 x 31 cm) | |
Ideal for applying thinners and solvents as well as removing lubricants, grease, soil and adhesives, general purpose |