missing translation for 'onlineSavingsMsg'
Learn More

Ansell Kimtech Pure™ A5 Cleanroom Non-Sterile Hoods

Non-sterile cleanroom hoods cover the head, neck, and shoulders to prevent contamination in the environment. Availble with or without ties.

$396.49 - $422.73

Specifications

Product Type Open Face
Size Universal
Material Polypropylene SMS Fabric
Color White
Latex Free Yes
View More Specs

Products 2
Catalog Number Mfr. No. Closure Type Price Quantity  
Catalog Number Mfr. No. Closure Type Price Quantity  
19-169-868
View Documents
Ansell
25796
Ties
Case of 100 Each for $422.73
Only null left
 
19-812-863
View Documents
Ansell
88849
None
Case of 100 Each for $396.49
Only null left
 
Description

Description

  • Non-sterile, white
  • Triple bagged, vacuum packed
  • Suitable for ISO Class 5 or higher cleanrooms; CE certified
  • Works with a variety of masks
  • Latex-free
  • Kimtech™ A5 Cleanroom Apparel is manufactured in an ISO-certified facility, clean-processes and individually packaged under cleanroom conditions
Specifications

Specifications

Open Face
Polypropylene SMS Fabric
Yes
Triple-bagged, Vacuum Packaged
ISO 5 to 8 cleanroom environments, compounding, mixing, filling, cleaning
Universal
White
Non-sterile
100/Cs.
Class 100 (ISO 5)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'