missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark Professional™ Scott™ Pro Foam Hair and Body Wash
Two products in one – a gentle body wash and a foaming shampoo. Luxury foam is protein-enriched with emollients and other skin conditioning agents. Foams nicely with pleasant fresh scent. Rinses clean without leaving a soapy residue. Great choice for health clubs, gyms, hospitality – anywhere where guests need to use your shower facilities.
Supplier: Kimberly-Clark Professional™ 91553
Description
- 1.0 L cassettes of Scott Pro Foam Body & Hair Wash; 6 cassettes/case
- Versatile (2-in-1) shampoo and body wash is a great choice where guests need to use the shower
- Rinses clean with no soapy residue
- Contains emollients and skin conditioning ingredients
- Use Scott Pro Foam Hair & Body Wash with a compatible Scott Pro dispenser for convenient, consistent dispensing; Itâs easy to refill when you buy in bulk
- Fresh, light scent
Specifications
Hair and Body Wash | |
Foam | |
5°C to 37°C | |
2.4 lb. |
Light Blue | |
5 to 6 | |
33.8 oz. | |
0.9 kg |