Learn More
Ansell KleenGuard™ A10 Bouffant Caps
KleenGuard™ A10 Bouffant Caps provide economical, disposable protection for workers exposed to dirt and grime in the workplace.
Supplier: Ansell 36920
Description
- Protective wear designed for entry-level protection from hazardous dust and feature an elastic opening
- Breathable fabric is made of lightweight spunbound material providing a cool, comfortable work environment
- Ideal choice for food processing, general maintenance and manufacturing
- Not made with natural rubber
- Elasticized opening
- A10 fabric is five layers of stacked barrier protection (SMMMS) against hazardous particles, offering economical protection
Specifications
Bouffant | |
Spunbond Polypropylene | |
36920 |
24 in. | |
Single-use | |
Food Processing, General Maintenance, and Manufacturing |