missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIAA0317 Monoclonal Antibody (K56023_20C3), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA555640
Description
Sequence of this protein is as follows: FDIIVLSEDE KNIVERNVST SGVSIYFEAY LYNATNCSST PWHLPPMHMT SSQRRPSTAV DEEDEDSPSE CHTPEKVKKP
KIAA0317 contains 1 filamin repeat and 1 HECT (E6AP-type E3 ubiquitin-protein ligase) domain. The exact function of KIAA0317 remains unknown.Specifications
KIAA0317 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
O15033 | |
AREL1 | |
Recombinant human AREL1 (immunogen range is 271-350). | |
50 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG2b |
Immunoprecipitation, Western Blot | |
K56023_20C3 | |
Unconjugated | |
AREL1 | |
apoptosis resistant E3 ubiquitin protein ligase 1; apoptosis-resistant E3 ubiquitin protein ligase 1; Apoptosis-resistant HECT-type E3 ubiquitin transferase 1; AREL1; fibrosis-inducing E3 ligase 1; FIEL1; KIAA0317; protein KIAA0317 | |
Mouse | |
Protein A | |
RUO | |
299197, 9870 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |