missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Khellin, 95%
Chemical Identifiers
CAS | 82-02-0 |
---|---|
Molecular Formula | C14H12O5 |
Molecular Weight (g/mol) | 260.25 |
InChI Key | HSMPDPBYAYSOBC-UHFFFAOYSA-N |
IUPAC Name | 4,9-dimethoxy-7-methyl-5H-furo[3,2-g]chromen-5-one |
SMILES | COC1=C2OC=CC2=C(OC)C2=C1OC(C)=CC2=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC461800050
|
Thermo Scientific™
461800050 |
5 g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC461800250
|
Thermo Scientific™
461800250 |
25 g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
82-02-0 | |
260.25 | |
4,9-dimethoxy-7-methyl-5H-furo[3,2-g]chromen-5-one |
C14H12O5 | |
HSMPDPBYAYSOBC-UHFFFAOYSA-N | |
COC1=C2OC=CC2=C(OC)C2=C1OC(C)=CC2=O |
Specifications
82-02-0 | |
C14H12O5 | |
Solubility aq. soln.: soluble. | |
COC1=C2OC=CC2=C(OC)C2=C1OC(C)=CC2=O | |
260.25 | |
Khellin |
95% | |
5 g | |
HSMPDPBYAYSOBC-UHFFFAOYSA-N | |
4,9-dimethoxy-7-methyl-5H-furo[3,2-g]chromen-5-one | |
260.24 |
Safety and Handling
GHS H Statement Toxic if swallowed.
GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
EINECSNumber : 201-392-8
RUO â Research Use Only