missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell KGA10 Coveralls, Elastic Wrists and Open Ankles
Light-duty coveralls offer protection against dirt and grime. Popular for agriculture, food processing, maintenance, labs, and dusty environments.
$171.20 - $171.20
Specifications
Product Type | Light-duty Coveralls |
---|---|
Gender | Unisex |
Material | Spunbond Polypropylene |
Color | White |
Disposable | Single-use |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
17-101-147
|
Ansell
67304 |
X-Large |
Case of 50 Each for $171.20
|
|
|||||
Description
- For non-hazardous particulate protection
- Lightweight and highly breathable spunbond fabric allows moisture to escape for maximum comfort
- Elastic wrist and open ankle design
- Not made with natural rubber latex or silicone
- Recyclable with the RightCycle™ Program
Specifications
Light-duty Coveralls | |
Spunbond Polypropylene | |
Single-use | |
Elastic | |
Unhooded | |
Stitched | |
Dust | |
CE Category I PPE | |
KleenGuard™ KGA10 |
Unisex | |
White | |
Open | |
Collared | |
Front Zipper | |
Non-sterile | |
ANSI/ISEA 101-2014 | |
Yes | |
Agriculture, Food Processing, General Maintenance, Laboratories |