missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Indoor Drum Storage Cabinet

Justrite's Indoor Drum Storage Cabinet features a removable, adjustable, half-depth shelf with enough headspace to accommodate a drum with a pump or funnel. This means the cabinet can be used as a pumping or waste accumulation station , storage for Class

Supplier:  Justrite™ 899100

Catalog No. 29-636-2128


Only null left
Explore available promotions

Description

Description

  • Double-wall 18-gauge steel with 1.5 in. (3.8cm) insulating air space
  • Three-point self-latching manual doors with double key set
  • Dual 2 in. (5cm) vents with flame arresters
  • Spill-catcher shelf adjustable on 2.25 in. (5.7cm) centers
  • Adjustable leveling feet
  • High-visibility warning label
  • 2 in. (5cm) leakproof sill to capture spills
  • Welded for superior strength
  • Powder-paint finish for maximum durability and chemical resistance

Order Info

Two capacities: hold either one or two 55-gal. (208L) drums.

Certifications

FM approved (except 17-150B).

Compliance

All meet NFPA and OSHA requirements.

Specifications

Specifications

142.2 cm
65 in.
Half-depth shelf, installed pumps, drum support, level adjusting feet and hazardous warning label
34 in.
30.34 x 56 x 60.5 in. (77.1 x 142.2 x 153.7 cm)
610 lb.
1
60.5 in.
Vertical drum storage cabinet
59 in.
Yellow
Manual-close
NFPA, OSHA
77.1 cm
Steel (CR)
277 kg
30.34 in.
86.4 cm
110 gal.
56 in.
153.7 cm
Flammables
149.9 cm
165.1 cm
2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions