missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ EcoPolyBlend™ Drum Management System
Features a space-saving stackable design
$600.74 - $2060.82
Specifications
Certifications/Compliance | SPCC, EPA, NFPA, OSHA |
---|---|
Color | Black |
Material | Polyethylene |
For Use With (Application) | Efficient storage, positive spill control, liquid dispensing |
Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
19-413-815
|
Justrite™
28667 |
Drum Management Base Module |
Each for $2,060.82
|
|
|||||
19-413-820
|
Justrite™
28671 |
Dispensing Shelf |
Each for $600.74
|
|
|||||
19-413-816
|
Justrite™
28669 |
Drum Management Stack Module |
Each for $1,354.53
|
|
|||||
Description
100% molded polyethylene construction provides maximum chemical resistance, preventing corrosion, deterioration and leaks
- Stores up to four 110 and/or 200L (30 and/or 55 gal.) drums
- Stores drums of Class 1B or 1C flammables and combustibles or nonhazardous liquids
- Consists of three parts: a base module, stack module and dispensing shelf
- Molded-in drum lock-channels hold drums in place
Base Module
- Holds two drums with a 1388kg (3060 lb.) total load capacity
- Easily relocates with an empty forklift
- 250L (66 gal.) open-view containment sump makes it easy to spot any spills for a quick cleanup
- Holds two additional drums to double the system capacity for a load of 680kg (1500 lb.)
- When storing both 110 and 200L (30 and 55-gal.) drums, the smaller drums should be placed on the top of the stack module
- Mounts securely to the stack module for convenient dispensing from the upper level
- Has a load capacity of 27 kg (60 lb.)
Compliance
EPA, NFPA, OSHA, SPCC
Specifications
SPCC, EPA, NFPA, OSHA | |
Black | |
Polyethylene | |
Efficient storage, positive spill control, liquid dispensing |