Learn More
Justrite™ EN Safety Cabinets for Flammables
Provides 30 minutes of fire resistance with safe storage and segregation of flammables
Supplier:  Justrite™ JMC22603
 View more versions of this product
Catalog No. 29-636-6282
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Attractive two-toned yellow cabinet
  • Constructed with body of melamine resin and flame retardant special plates; Steel door front covered by textured, durable powder-coat finish
  • Provides 30 minutes of content protection and extended evacuation time Hybrid style door open or close manually and include damper that adjusts force needed to move doors
  • Under fire conditions, intumescent strips expand and hermetically seal all gaps to reduce heat entering cabinet and built-in fusible link system releases to allow automatic closure of doors at 122°F (50°C)
  • Powder-coat finish steel shelves adjust on 32mm (1.25 in.) increments with load capacity of 75kg (165 lb)
  • Shelves with high ledge prevent small containers from sliding off
  • Removable bottom sump with detachable perforated insert features three-sided lip seal to capture leaks or spills from shelves at bottom of sump, keeping containers in sump clean and dry
Certifications
Tested and GS certified by MPA Dresden
Compliance
EN 14470-1 / EN 14727 compliant and meet NFPA 30, NFPA 1, and OSHA standards
Specifications
Specifications
30 minutes fire resistance; Melamine resin, flame-resistant special plates
45 gal.
Melamine Resin
2
Product Suggestions
Product Suggestions