Learn More
Johnson Scintillation Probes
Supplier: Johnson GP200
Specifications
approx. 2100 cpm/mR/hr. (Cs-137 gamma) | |
C-14 -- 2%, SrY-90 -- 10%, Tc-99 -- 7%, I-125 -- 0.1% | |
Active: 6 sq cm; Open: 5 sq cm |
BNC | |
General purpose detector sensitive to most alpha, beta, and gamma radiation | |
C-14, SrY-90, Tc-99 and Cs-137 |