Learn More
Fisherbrand™ Isotemp Series General Purpose Ultra-Low Freezer
Isotemp™ ULT Freezers combine reliability and performance with cost-effective operation and innovative features.
$8208.28 - $8314.60
Specifications
For Use With (Equipment) Isotemp Series Upright ULT Freezers
View More Specs
Products 2
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
FCOPLC
View Documents
Fisherbrand™
FCOPLC
CO2 Backup System
Each for $8,314.60
Only null left
FLNPLC
View Documents
Fisherbrand™
FLNPLC
LN2 Backup System
Each for $8,208.28
Only null left
Description
Description
  • Isotemp™ Ultra-Low-Temperature Freezers combine reliability and performance with cost-effective operation and innovative features.
  • They are built using an advanced technology platform, refrigeration, microprocessor controls, and high-quality construction.
  • Performance
  • Maximum temperature recovery: Reduced/low peak temperature variation and fast open-door recovery
  • More comfortable lab environment: Low noise levels (like your refrigerator at home2)
  • Operational efficiency: Lower energy consumption and HVAC costs
  • User Friendly
  • Single-handed door operation
  • Five inner compartments (with doors)
  • Large temperature display with touch control
  • Centralized settings and controls with battery backup
  • Alarms, security passcode, and padlock-compatible doors
  • Easy access to the filter/grill assembly (cleaning and maintenance)
  • Sustainability
  • Manufactured in award winning zero-waste to landfill facility
  • Natural, green hydrocarbon refrigerants improve performance and increase reliability with low GWP and zero ODP
  • Environmentally friendly: Water-blown foam insulation; compliant with EPA SNAP and EU F-Gas regulations
  • Additional features
  • Standard RS-485, 4 - 20 mA analog outputs and dry contacts included
  • Battery backup for control panel (power interruptions)
  • Four capacities; Universal voltage
  • No setup required
  • Warranty
  • Five years parts and labor
Specifications
Specifications
Isotemp Series Upright ULT Freezers
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'