missing translation for 'onlineSavingsMsg'
Learn More

Isoproturon 98.0+%, TCI America™

$208.78 - $630.05

Chemical Identifiers

CAS 34123-59-6
Molecular Formula C12H18N2O
Molecular Weight (g/mol) 206.289
MDL Number MFCD00078684
InChI Key PUIYMUZLKQOUOZ-UHFFFAOYSA-N
Synonym isoproturon, graminon, 3-4-isopropylphenyl-1,1-dimethylurea, tolkan, arelon, belgran, arelon r, nocilon, ipu stefes, ip-flo
PubChem CID 36679
ChEBI CHEBI:6049
IUPAC Name 1,1-dimethyl-3-(4-propan-2-ylphenyl)urea
SMILES CC(C)C1=CC=C(C=C1)NC(=O)N(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I08435G
View Documents
TCI America
I08435G
5 g
Each for $208.78
Only null left
 
I084325G
View Documents
TCI America
I084325G
25 g
Each for $630.05
Only null left
 
Description

Description

Specifications

Chemical Identifiers

34123-59-6
206.289
PUIYMUZLKQOUOZ-UHFFFAOYSA-N
36679
1,1-dimethyl-3-(4-propan-2-ylphenyl)urea
C12H18N2O
MFCD00078684
isoproturon, graminon, 3-4-isopropylphenyl-1,1-dimethylurea, tolkan, arelon, belgran, arelon r, nocilon, ipu stefes, ip-flo
CHEBI:6049
CC(C)C1=CC=C(C=C1)NC(=O)N(C)C

Specifications

34123-59-6
White
MFCD00078684
3077
PUIYMUZLKQOUOZ-UHFFFAOYSA-N
1,1-dimethyl-3-(4-propan-2-ylphenyl)urea
36679
206.29
Crystalline Powder
158°C
C12H18N2O
5 g
isoproturon, graminon, 3-4-isopropylphenyl-1,1-dimethylurea, tolkan, arelon, belgran, arelon r, nocilon, ipu stefes, ip-flo
CC(C)C1=CC=C(C=C1)NC(=O)N(C)C
206.289
CHEBI:6049
≥98.0% (HPLC,N)
Isoproturon
Videos
Safety and Handling

Safety and Handling

RTECSNumber : YT0170000

TSCA : No

SDS
Documents

Documents