missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Rubbing Alcohol, USP, 68-72%, Spectrum™ Chemical

IS120, 67-63-0, C3H8O

Supplier:  Spectrum Chemical Mfg Cor IS1204LTGL

 View more versions of this product

Catalog No. 18603567


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Isopropyl Rubbing Alcohol, 70 Percent, USP , is a colorless and flammable chemical compound also known as Rubbing Alcohol. It is a cooling and soothing liquid for external applications to minimize the risk of infection and is not suitable for internal consumption. Spectrum Chemical USP products, including all USP alcohol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Specifications

67-63-0,7732-18-5
4 L
0.872-0.883
UN1219
CC(C)O
60.10
USP
70%,30%
Amber Glass Bottle
C3H8O
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
68 to 72%

Chemical Identifiers

67-63-0
60.10
propan-2-ol
C3H8O
KFZMGEQAYNKOFK-UHFFFAOYSA-N
CC(C)O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions