missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Alcohol, FCC, 99%, Spectrum™ Chemical
I1056, 67-63-0, C3H8O
Supplier: Spectrum Chemical Mfg Cor I10564LTPL
Description
Spectrum™ Chemical Isopropyl Alcohol, 99 Percent, FCC is used to sanitize food processing equipment. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
67-63-0 | |
100% | |
Poly Bottle | |
C3H8O | |
0.2% | |
CC(C)O | |
60.10 | |
FCC |
10 mg/kg | |
4 L | |
0.784 | |
UN1219 | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
propan-2-ol | |
99.7% |
Chemical Identifiers
67-63-0 | |
60.10 | |
propan-2-ol |
C3H8O | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
CC(C)O |