missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, 99%, EP, BP, JP, USP, 99%, Spectrum™ Chemical

IS115, 67-63-0, C3H8O

Supplier:  Spectrum Chemical Mfg Cor IS115500MLGLCS6

 View more versions of this product

Catalog No. 18603526


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Isopropyl Alcohol, 99 Percent, USP, EP, BP, JP has many uses, including as a pharmaceutical raw material for preparing USP Rubbing Alcohol for relief of muscle stiffness and an infection preventative for minor scrapes and burns. All Spectrum Chemical USP, EP, BP, JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Specifications

67-63-0
Clear
Amber Glass Bottle
0.785-0.788
UN1219
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
99%
100%
6 x 500 mL
1.376-1.378
C3H8O
0.5%
CC(C)O
60.10

Chemical Identifiers

67-63-0
60.10
propan-2-ol
C3H8O
KFZMGEQAYNKOFK-UHFFFAOYSA-N
CC(C)O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions