Learn More
ISOPROPYL ALCOHOL OMNI 4L
Supplier: Technidata PX1834P1
Specifications
Isopropyl Alcohol | |
Titrable acid: 0.2µeq/g | |
-89.5°C | |
.786 g/cc at 20°C | |
425°C | |
Glas bottle (Pre packed) | |
4 L | |
1ppm | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
propan-2-ol | |
3776 | |
60.10 | |
99.9% | |
Liquid |
67-63-0 | |
82.4°C (1013 hPa) | |
Colorless | |
12°C | |
Alcohol-like | |
C3H8O | |
UN 1219 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CC(C)O | |
60.096 | |
CHEBI:17824 | |
43 hPa (20°C) | |
OmniSolv™ |
Chemical Identifiers
67-63-0 | |
60.096 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CHEBI:17824 | |
CC(C)O |
C3H8O | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
3776 | |
propan-2-ol |
Safety and Handling
Hazard Statements H225 Highly flammable liquid and vapor. H319 Causes serious eye irritation. H336 May cause drowsiness or dizziness
Precautionary Statements P210 Keep away from heat/sparks/open flames/hot surfaces. - No smoking. P233 Keep container tightly closed. P240 Ground/bond container and receiving equipment. P305 + P351 + P338 IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. P403 + P235 Store in a well-ventilated place. Keep cool
Danger
EINECSNumber : 200-661-7
RTECSNumber : NT8050000
Recommended Storage : Store at room temperature.