missing translation for 'onlineSavingsMsg'
Learn More

Isopropanol/Water, 91:9, Reagents

Isopropanol/Water, 91:9, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

$198.30 - $9020.20

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym Isopropyl Alcohol, 2-Propanol, Propan-2-ol
IUPAC Name propan-2-ol
SMILES CC(C)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01-334-897
View Documents
Labchem Inc
CI298100200E
200 L
Each for $9,020.20
Only null left
 
01-334-896
View Documents
Labchem Inc
CI2981004A
4 L
Each for $579.47
Only null left
 
01-334-895
View Documents
Labchem Inc
CI298100500A
500 mL
Each for $198.30
Only null left
 
Description

Description

Specifications

Chemical Identifiers

67-63-0
60.10
Isopropyl Alcohol, 2-Propanol, Propan-2-ol
CC(C)O
C3H8O
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol

Specifications

67-63-0,7732-18-5
82°C
Poly Drum
0.8159
Isopropyl Alcohol, 2-Propanol, Propan-2-ol
CC(C)O
60.10
Reagent
Colorless
200 L
Liquid
C3H8O
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
91%
Isopropanol/Water, 91:9
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'