missing translation for 'onlineSavingsMsg'
Learn More

Isobutyric Acid 99.0+%, TCI America™

$35.41 - $54.06

Chemical Identifiers

CAS 79-31-2
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00002658
InChI Key KQNPFQTWMSNSAP-UHFFFAOYSA-N
Synonym isobutyric acid, isobutanoic acid, 2-methylpropionic acid, propanoic acid, 2-methyl, dimethylacetic acid, isopropylformic acid, iso-butyric acid, isobutyrate, cenex rp b2, acetic acid, dimethyl
PubChem CID 6590
ChEBI CHEBI:16135
IUPAC Name 2-methylpropanoic acid
SMILES CC(C)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I010325ML
View Documents
TCI America
I010325ML
25 mL
Each for $35.41
Only null left
 
I0103500ML
View Documents
TCI America
I0103500ML
500 mL
Each for $54.06
Only null left
 
Description

Description

Specifications

Chemical Identifiers

79-31-2
88.11
KQNPFQTWMSNSAP-UHFFFAOYSA-N
6590
2-methylpropanoic acid
C4H8O2
MFCD00002658
isobutyric acid, isobutanoic acid, 2-methylpropionic acid, propanoic acid, 2-methyl, dimethylacetic acid, isopropylformic acid, iso-butyric acid, isobutyrate, cenex rp b2, acetic acid, dimethyl
CHEBI:16135
CC(C)C(O)=O

Specifications

-47°C
155°C
C4H8O2
2529
KQNPFQTWMSNSAP-UHFFFAOYSA-N
2-methylpropanoic acid
6590
88.11
25 mL
Isobutyric Acid
79-31-2
Colorless
MFCD00002658
isobutyric acid, isobutanoic acid, 2-methylpropionic acid, propanoic acid, 2-methyl, dimethylacetic acid, isopropylformic acid, iso-butyric acid, isobutyrate, cenex rp b2, acetic acid, dimethyl
CC(C)C(O)=O
88.11
CHEBI:16135
≥99.0% (GC)
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0608

RTECSNumber : NQ4375000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'