missing translation for 'onlineSavingsMsg'
Learn More

Isobutylamine Hydrobromide 98.0+%, TCI America™

$76.43 - $229.29

Chemical Identifiers

CAS 74098-36-5
Molecular Formula C4H12BrN
Molecular Weight (g/mol) 154.05
MDL Number MFCD29089367
InChI Key RFYSBVUZWGEPBE-UHFFFAOYSA-N
Synonym Isobutylammonium Bromide
PubChem CID 85987475
IUPAC Name 2-methylpropan-1-amine hydrobromide
SMILES Br.CC(C)CN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I10071G
View Documents
TCI America
I10071G
1 g
Each for $76.43
Only null left
 
I10075G
View Documents
TCI America
I10075G
5 g
Each for $229.29
Only null left
 
Description

Description

Specifications

Chemical Identifiers

74098-36-5
154.05
RFYSBVUZWGEPBE-UHFFFAOYSA-N
85987475
Br.CC(C)CN
C4H12BrN
MFCD29089367
Isobutylammonium Bromide
2-methylpropan-1-amine hydrobromide

Specifications

74098-36-5
White
MFCD29089367
Isobutylammonium Bromide
Br.CC(C)CN
154.05
154.05
Crystalline Powder
182°C
C4H12BrN
1 g
RFYSBVUZWGEPBE-UHFFFAOYSA-N
2-methylpropan-1-amine hydrobromide
85987475
≥98.0% (N,T)
Isobutylamine Hydrobromide
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'