missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iso-Octane, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.0%, Honeywell Burdick & Jackson
$30469.40 - $30469.40
Chemical Identifiers
CAS | 540-84-1 |
---|---|
Molecular Formula | C8H18 |
Molecular Weight (g/mol) | 114.232 |
InChI Key | NHTMVDHEPJAVLT-UHFFFAOYSA-N |
Synonym | isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane |
PubChem CID | 10907 |
ChEBI | CHEBI:62805 |
IUPAC Name | 2,2,4-trimethylpentane |
SMILES | CC(C)CC(C)(C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
6000872
|
Honeywell Chemicals
PP362204 |
204 L | Metal Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
540-84-1 | |
114.232 | |
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane | |
CHEBI:62805 | |
CC(C)CC(C)(C)C |
C8H18 | |
NHTMVDHEPJAVLT-UHFFFAOYSA-N | |
10907 | |
2,2,4-trimethylpentane |
Specifications
540-84-1 | |
0.69 g/cm3 | |
204 L | |
Metal Drum | |
UN1262 | |
1696876 | |
0.02% in Water at 20°C | |
CC(C)CC(C)(C)C | |
114.232 | |
CHEBI:62805 | |
>99% | |
Iso-Octane, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.0% |
100.0000% | |
99.24°C | |
−8°C | |
C8H18 | |
0.01% max. | |
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane | |
NHTMVDHEPJAVLT-UHFFFAOYSA-N | |
2,2,4-trimethylpentane | |
10907 | |
114.23 | |
ACS Reagent | |
by GC Analysis |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Octanes