missing translation for 'onlineSavingsMsg'
Learn More

Iso-Octane, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.0%, Honeywell Burdick & Jackson

$1392.50 - $1392.50

Chemical Identifiers

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
PubChem CID 10907
ChEBI CHEBI:62805
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
6000868
View Documents
Honeywell-Burdick and Jackson
3621L
6 x 1 L Glass Bottle
Case of 6 Each for $1,392.50
Only null left
 
Description

Description

Specifications

Chemical Identifiers

540-84-1
114.232
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
CHEBI:62805
CC(C)CC(C)(C)C
C8H18
NHTMVDHEPJAVLT-UHFFFAOYSA-N
10907
2,2,4-trimethylpentane

Specifications

540-84-1
100.0000%
99.24°C
−8°C
C8H18
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
CC(C)CC(C)(C)C
114.232
CHEBI:62805
>99%
Iso-Octane, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.0%
0.1 AU max. at 225 nm, 0.02 AU max. at 250 nm, 0.005 AU max. at 300 nm
0.69 g/cm3
6 x 1 L
Glass Bottle
UN1262
NHTMVDHEPJAVLT-UHFFFAOYSA-N
2,2,4-trimethylpentane
10907
114.23
HPLC
Videos
Safety and Handling

Safety and Handling

ShelfLife : 547 days from date of manufacture

DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Octanes

SDS
missing translation for 'documents'

missing translation for 'documents'