missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ishikawa's Reagent 92.0+%, TCI America™
$147.40 - $432.76
Chemical Identifiers
CAS | 309-88-6 |
---|---|
Molecular Formula | C7H11F6N |
Molecular Weight (g/mol) | 223.162 |
MDL Number | MFCD00060264 |
InChI Key | BNTFCVMJHBNJAR-UHFFFAOYSA-N |
Synonym | n,n-diethyl-1,1,2,3,3,3-hexafluoropropylamine, n,n-diethyl 1,1,2,3,3,3-hexafluoropropylamine, diethyl 1,1,2,3,3,3-hexafluoropropyl amine, 1,1,2,3,3,3-hexafluoro-1-diethylaminopropane, n,n-diethyl-1,1,2,3,3,3-hexafluoro-1-propanamine, 1-propanamine, n,n-diethyl-1,1,2,3,3,3-hexafluoro, n,n-diethyl-1,1,2,3,3,3-hexafluoropropyl amine, ishikawa s reagent, acmc-20ak9c, ishikawa's reagant |
PubChem CID | 136149 |
IUPAC Name | N,N-diethyl-1,1,2,3,3,3-hexafluoropropan-1-amine |
SMILES | CCN(CC)C(C(C(F)(F)F)F)(F)F |
Chemical Identifiers
309-88-6 | |
223.162 | |
BNTFCVMJHBNJAR-UHFFFAOYSA-N | |
136149 | |
CCN(CC)C(C(C(F)(F)F)F)(F)F |
C7H11F6N | |
MFCD00060264 | |
n,n-diethyl-1,1,2,3,3,3-hexafluoropropylamine, n,n-diethyl 1,1,2,3,3,3-hexafluoropropylamine, diethyl 1,1,2,3,3,3-hexafluoropropyl amine, 1,1,2,3,3,3-hexafluoro-1-diethylaminopropane, n,n-diethyl-1,1,2,3,3,3-hexafluoro-1-propanamine, 1-propanamine, n,n-diethyl-1,1,2,3,3,3-hexafluoro, n,n-diethyl-1,1,2,3,3,3-hexafluoropropyl amine, ishikawa s reagent, acmc-20ak9c, ishikawa's reagant | |
N,N-diethyl-1,1,2,3,3,3-hexafluoropropan-1-amine |
Specifications
309-88-6 | |
54°C | |
MFCD00060264 | |
2920 | |
BNTFCVMJHBNJAR-UHFFFAOYSA-N | |
N,N-diethyl-1,1,2,3,3,3-hexafluoropropan-1-amine | |
136149 | |
≥92.0% (T) | |
Ishikawa′s Reagent [Fluorinating Reagent] |
Yellow | |
C7H11F6N | |
25 g | |
n,n-diethyl-1,1,2,3,3,3-hexafluoropropylamine, n,n-diethyl 1,1,2,3,3,3-hexafluoropropylamine, diethyl 1,1,2,3,3,3-hexafluoropropyl amine, 1,1,2,3,3,3-hexafluoro-1-diethylaminopropane, n,n-diethyl-1,1,2,3,3,3-hexafluoro-1-propanamine, 1-propanamine, n,n-diethyl-1,1,2,3,3,3-hexafluoro, n,n-diethyl-1,1,2,3,3,3-hexafluoropropyl amine, ishikawa s reagent, acmc-20ak9c, ishikawa's reagant | |
CCN(CC)C(C(C(F)(F)F)F)(F)F | |
223.162 | |
223.16 | |
Liquid |
Safety and Handling
TSCA : No