missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron Metal, Reduced, Powder, 96%, Spectrum™ Chemical
$364.70 - $11111.60
Chemical Identifiers
CAS | 7439-89-6 |
---|---|
Molecular Formula | Fe |
Molecular Weight (g/mol) | 55.85 |
MDL Number | MFCD00010999 |
InChI Key | XEEYBQQBJWHFJM-UHFFFAOYSA-N |
IUPAC Name | iron |
SMILES | [Fe] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18611090
|
Spectrum Chemical Mfg Cor
I1035125GM |
125 g | Poly Bottle |
Each for $364.70
|
|
||||
18611092
|
Spectrum Chemical Mfg Cor
I1035500GM |
500 g | Poly Bottle |
Each for $781.07
|
|
||||
18611094
|
Spectrum Chemical Mfg Cor
I103525KG |
2.5 kg | Poly Bottle |
Each for $2,058.23
|
|
||||
18611091
|
Spectrum Chemical Mfg Cor
I103512KG |
12 kg | Poly Pail |
Each for $4,641.73
|
|
||||
18611093
|
Spectrum Chemical Mfg Cor
I103550KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
7439-89-6 | |
55.85 | |
XEEYBQQBJWHFJM-UHFFFAOYSA-N | |
[Fe] |
Fe | |
MFCD00010999 | |
iron |
Specifications
7439-89-6 | |
Fe | |
125 g | |
[Fe] | |
55.85 | |
Reagent |
1 | |
MFCD00010999 | |
XEEYBQQBJWHFJM-UHFFFAOYSA-N | |
iron | |
96% | |
Poly Bottle |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.