missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(II) chloride tetrahydrate, Honeywell Fluka™
Puriss. p.a.,≥99.0% (RT)
$762.95 - $762.95
Chemical Identifiers
CAS | 13478-10-9 |
---|---|
Molecular Formula | Cl2FeH8O4 |
Molecular Weight (g/mol) | 198.805 |
MDL Number | MFCD00149709 |
InChI Key | WSSMOXHYUFMBLS-UHFFFAOYSA-L |
Synonym | ferrous chloride tetrahydrate, iron ii chloride tetrahydrate, iron dichloride tetrahydrate, cl2fe.4h2o, fecl2.4h2o, iron chloride, tetrahydrate, dichloroiron-water 1/4, iron ii chloride tetra hydrate, ae>> nciu eae r masculineiii |
PubChem CID | 16211588 |
ChEBI | CHEBI:86249 |
IUPAC Name | dichloroiron;tetrahydrate |
SMILES | O.O.O.O.Cl[Fe]Cl |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
13478-10-9 | |
198.805 | |
WSSMOXHYUFMBLS-UHFFFAOYSA-L | |
16211588 | |
dichloroiron;tetrahydrate |
Cl2FeH8O4 | |
MFCD00149709 | |
ferrous chloride tetrahydrate, iron ii chloride tetrahydrate, iron dichloride tetrahydrate, cl2fe.4h2o, fecl2.4h2o, iron chloride, tetrahydrate, dichloroiron-water 1/4, iron ii chloride tetra hydrate, ae>> nciu eae r masculineiii | |
CHEBI:86249 | |
O.O.O.O.Cl[Fe]Cl |
Specifications
13478-10-9 | |
Cl2FeH8O4 | |
MFCD00149709 | |
ferrous chloride tetrahydrate, iron ii chloride tetrahydrate, iron dichloride tetrahydrate, cl2fe.4h2o, fecl2.4h2o, iron chloride, tetrahydrate, dichloroiron-water 1/4, iron ii chloride tetra hydrate, ae>> nciu eae r masculineiii | |
O.O.O.O.Cl[Fe]Cl | |
198.805 | |
CHEBI:86249 | |
≥99% (RT) | |
Iron(II) chloride tetrahydrate |
1 kg | |
FeCl2 · 4H2O | |
UN3260 | |
WSSMOXHYUFMBLS-UHFFFAOYSA-L | |
dichloroiron;tetrahydrate | |
16211588 | |
198.81g/mol | |
10 mmHg (693°C) |
Safety and Handling
P280-P305 + P351 + P338-P310
RTECSNumber : NO5600000