missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ iQ Series™ Men's Lightweight Navy Flame Resistant Pant
iQ Series™ Men's Lightweight Navy Flame Resistant Pant is stylishly designed to keep you cool and in command all day long.
$236.99 - $236.99
Specifications
Gender | Men's |
---|---|
Material | Aramid, Modacrylic, Lyocell |
Color | Navy |
Seam Type | Stitched |
Closure Type | Concealed Button |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
17-204-071
|
Bulwark™
QP18NV2837U |
28 x 37 in. |
Each for $236.99
|
|
|||||
17-204-046
|
Bulwark™
QP18NV3037U |
30 x 37 in. |
Each for $236.99
|
|
|||||
17-204-452
|
Bulwark™
QP18NV3437U |
34 x 37 in. |
Each for $236.99
|
|
|||||
17-204-290
|
Bulwark™
QP18NV3637U |
36 x 37 in. |
Each for $236.99
|
|
|||||
17-204-458
|
Bulwark™
QP18NV3837U |
38 x 37 in. |
Each for $236.99
|
|
|||||
17-204-395
|
Bulwark™
QP18NV4037U |
40 x 37 in. |
Each for $236.99
|
|
|||||
17-204-447
|
Bulwark™
QP18NV4237U |
42 x 37 in. |
Each for $236.99
|
|
|||||
17-204-455
|
Bulwark™
QP18NV4437U |
44 x 37 in. |
Each for $236.99
|
|
|||||
17-203-953
|
Bulwark™
QP18NV4637U |
46 x 37 in. |
Each for $236.99
|
|
|||||
17-204-037
|
Bulwark™
QP18NV4837U |
48 x 37 in. |
Each for $236.99
|
|
|||||
17-204-038
|
Bulwark™
QP18NV5037U |
50 x 37 in. |
Each for $236.99
|
|
|||||
17-204-080
|
Bulwark™
QP18NV5237U |
52 x 37 in. |
Each for $236.99
|
|
|||||
17-204-125
|
Bulwark™
QP18NV5437U |
54 x 37 in. |
Each for $236.99
|
|
|||||
17-204-147
|
Bulwark™
QP18NV5637U |
56 x 37 in. |
Each for $236.99
|
|
|||||
Description
- Let our work pant do a little more of the lifting.
- The Bulwark™ iQ Series™ Men's Lightweight Comfort Pant is stylishly designed to keep you cool and in command all day long.
- This modern, straight-fit jean style work pant features a one-piece waistband with wide belt loops, scoop front pockets, reinforced cargo pocket with snap closure on the left and patch hip pockets, a utility pocket on the right side plus triple needle out and inseam for extra durability.
- Certifications Level 1: ASTM F2302, NFPA 2112, NFPA 70E
- Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
- Hazard: Arc Flash, Flash Fire
- Number of Pockets: 6
- Primary Closure: Concealed Button
- Wash Care: Industrial Laundry - Light Soil
- Fabric: Body: 6.5 oz. (220 g/m&superscript_2;) TenCate Evolv™ Flame Resistant
- Blend: Body: 50% Aramid / 25% Modacrylic / 25% Lyocell
- Pocket: Two scoop front pockets, Two patch hip pockets, One cargo pocket with snap closure on left leg and one utility pocket on right leg
- Features: Triple-needle felled main seams, Reinforced hip pockets and cargo pocket, Jean style cut with one-piece waistband and concealed button closure
- Product Collection: iQ Series Comfort
- Protection: ATPV Arc Rating: 8.4
- Country of Origin: Imported, Made in USA Fabric
- Gender: Male
- Brand: Bulwark™
Specifications
Men's | |
Navy | |
Concealed Button | |
Category 2 | |
ASTM F2302, NFPA 2112, NFPA 70E |
Aramid, Modacrylic, Lyocell | |
Stitched | |
Reusable | |
8.4 |