missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine Monochloride Solution WIJS AOAC, Reagents
Supplier: Labchem Inc C7218100500C
Alert:
DEA LIST I
Description
- Iodine Monochloride Solution, WIJS
- For Determining Iodine Absorption Number of Fat and Oils
- Appearance: Brown liquid
- Specific Gravity: 1.05
- Boiling Point: 118°C
- Brand: Reagents
- Iodine/Chlorine Ratio Spec: 1.05 to 1.15
- Total Halogen Content (vs Sodium Thiosulfate/Starch) Spec: 0.189 to 0.193 N at 20°C
Specifications
Iodine Monochloride Solution, WIJS | |
7553-56-2,64-19-7,7790-99-0 | |
500 mL | |
100 | |
Safety Coated, Amber Glass Bottle |
Blend | |
Liquid or Solid | |
Blend | |
Laboratory | |
Amber Glass Bottle |