missing translation for 'onlineSavingsMsg'
Learn More

Iodine 0.4 Normal DEA List I, Reagents

Supplier:  Labchem Inc CI10200020E7

Catalog No. 361059166


Only null left
Explore available promotions

Alert:

DEA LIST I


Description

Description

  • Iodine 0.4 N
  • DEA List I
  • Appearance: Dark Brown Liquid
  • Size: 20 L
  • Specific Gravity: 1.15
  • Brand: Reagents
  • NIST Traceable
  • Prepared with ACS Grade Components and ASTM Type II Water
  • Assay (vs. Sodium Thiosulfate/Starch) Spec: 0.38 to 0.42 N
Specifications

Specifications

Liquid or Solid
Blend
Laboratory
20 L
0.40 N

Chemical Identifiers

Blend
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions