missing translation for 'onlineSavingsMsg'
Learn More

Iodine 0.356 Normal DEA List I, Reagents

Supplier:  Labchem Inc CI1010004C

Catalog No. 361059146


Only null left
Explore available promotions

Alert:

DEA LIST I


Description

Description

  • Iodine 0.356 N
  • DEA List I
  • Appearance: Dark Brown Liquid
  • Size: 4 L
  • Specific Gravity: 1.15
  • Brand: Reagents
  • NIST Traceable
  • Prepared with ACS Grade Components and ASTM Type II Water
Specifications

Specifications

Liquid or Solid
Blend
Laboratory
4 L
0.356 N

Chemical Identifiers

Blend
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions