missing translation for 'onlineSavingsMsg'
Learn More

Iodine 0.315 Normal DEA List I, Reagents

Supplier:  Labchem Inc CI1004001C

 View more versions of this product

Catalog No. 361059171


Only null left
Explore available promotions

Alert:

DEA LIST I


Description

Description

  • Iodine 0.315 N
  • DEA List I
  • Appearance: Dark Brown Liquid
  • Specific Gravity: 1.15
  • Brand: Reagents
  • NIST Traceable
  • Prepared with ACS Grade Components and ASTM Type II Water
  • Assay (vs. Sodium Thiosulfate/Starch) Spec: 0.3145 to 0.3155 N
Specifications

Specifications

Liquid or Solid
Blend
Laboratory
1 L
0.315 N

Chemical Identifiers

Blend
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions