missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine 0.3125 Normal DEA List I, Reagents
Supplier: Labchem Inc CI10060025C
Alert:
DEA LIST I
Description
- Iodine 0.3125 N
- DEA List I
- Appearance: Dark Brown Liquid
- Specific Gravity: 1.15
- Brand: Reagents
- NIST Traceable
- Prepared with ACS Grade Components and ASTM Type II Water
Specifications
Liquid or Solid | |
Blend | |
Laboratory |
2.5 L | |
0.3125 N |
Chemical Identifiers
Blend |