missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine 0.250 Normal DEA List I, Reagents
Supplier: Labchem Inc CI2024001C
Alert:
DEA LIST I
Description
- Iodine, 0.250 N
- DEA List I
- Appearance: Dark Brown Liquid
- Size: 1 L
- Specific Gravity: 1.11
- Brand: Reagents
- NIST Traceable
- Prepared with ACS Grade Components and ASTM Type II Water
Specifications
Liquid or Solid | |
Blend | |
Laboratory |
1 L | |
0.250 N |
Chemical Identifiers
Blend |