missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine, 0.00500 Normal, Reagents
Iodine, 0.00500 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CI201300500C
Specifications
100°C | |
Dark Brown | |
Liquid | |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
Reagent | |
1.0025 |
Iodine, 0.00500 Normal | |
7732-18-5,7681-11-0 | |
500 mL | |
Diiodine solution | |
O | |
18.02 | |
Glass Bottle |
Chemical Identifiers
7732-18-5 | |
18.02 | |
Diiodine solution | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water |