missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UltraPure™ SSC, 20X
For use in nucleic acid hybridizations and blot transfer applications
$95.25 - $267.00
Specifications
Content And Storage | Store at room temperature. |
---|---|
Concentration | 20 X |
Product Type | SSC |
Form | Liquid |
For Use With (Application) | Nucleic Acid Gel Electrophoresis, Blotting |
Description
UltraPure 20X SSC is a solution formulated and used in concentrations ranging from 0.2X to 20X, depending on the application.
- Supplied in 1 L plastic bottles/4 L/10 L stackable Cubitainer™ Box
- Sterile-filtered UltraPure 20X SSC contains 3.0 M NaCl and 0.3 M sodium citrate, at pH 7.0
- Performance and Quality Testing: No DNase or RNase activity detected
DNA & RNA Purification & Analysis, Gene Expression Analysis & Genotyping, Microarray Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis & Blotting, Southern Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Store at room temperature. | |
SSC | |
Nucleic Acid Gel Electrophoresis, Blotting | |
Room Temperature |
20 X | |
Liquid | |
UltraPure™ |
For Research Use Only. Not for use in diagnostic procedures.