missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ Installation Kits for Packed and Micropacked Columns
Installation kits for packed and micropacked columns for direct injections or valve applications.
$144.64 - $218.32
Catalog Number | Mfr. No. | For Use With | Diameter (Metric) Inner | For Use With (Application) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With | Diameter (Metric) Inner | For Use With (Application) | Price | Quantity | |||
06711395
![]() |
Restek™
RK21066 |
1 mm O.D. Columns | 1 mm | For Direct Injections |
Each for $218.32
|
|
|||
06711396
![]() |
Restek™
RK21067 |
2 mm I.D. Columns | 2 mm | For Valve Applications |
Each for $144.64
|
|
|||
06711394
![]() |
Restek™
RK21065 |
1 mm O.D. Columns | 1 mm | For Valve Applications |
Each for $171.32
|
|
|||
Description
Part No. 21065
- For micropacked columns
- For 1mm ID columns
- For valve applications
Part No. 21066
- For micropacked columns
- For 1mm ID columns
- For direct injections
Part No. 21067
- For packed columns
- For 2mm ID columns
- For valve applications
Specifications
Installation Kit |