missing translation for 'onlineSavingsMsg'
Learn More

Inositol, 97%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor IN14025KG

 View more versions of this product

Catalog No. 18611041


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

87-89-8
224° to 227°C
C6H12O6
CDAISMWEOUEBRE-UHFFFAOYSA-N
cyclohexane-1,2,3,4,5,6-hexol
0.97
1
Poly Pail
2.5 kg
OC1C(O)C(O)C(O)C(O)C1O
180.16
Reagent

Chemical Identifiers

87-89-8
180.16
cyclohexane-1,2,3,4,5,6-hexol
C6H12O6
CDAISMWEOUEBRE-UHFFFAOYSA-N
OC1C(O)C(O)C(O)C(O)C1O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.