missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Inorganic Carbon Standard, Reagents
Inorganic Carbon Standard, Reagent, use for Water analysis. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CC2058004C
Specifications
Inorganic Carbon Standard | |
7732-18-5,497-19-8 | |
Liquid | |
100°C | |
Completely miscible | |
4 L |
Glass Bottle | |
1.0045 | |
Colorless | |
Water Analyzer | |
Water Analysis | |
Reagent |