missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Inkjet Vinyl Label
Make an impact with this inkjet-printable vinyl label outfitted with a specially formulated topcoat that invigorates colors and label appearance.
Supplier: Brady™ J40C11252595
Description
- Flexible material withstands harsh elements, including liquids, alcohol, chemicals, and indoor fluorescent and UV lighting
- Topcoat also protects against smudges, smears and splotches
- An ink cartridge is required to print this material
Specifications
Not Autoclavable | |
None | |
Matte | |
Vinyl | |
Ink Jet | |
Non-sterile | |
J4000 Printer | |
1 Roll | |
-40°F to +140°F | |
0.0054 in. | |
100 ft. x 1.125 in. | |
30.48 m | |
28.58 mm |
Permanent Acrylic | |
White | |
Continuous | |
Single-side Printable | |
Labels | |
Facility Identification, General Identification, Hazard Communication Identification, Lean and 5S Labeling, Lockout Tagout, Pipe and Valve Marking, Warehouse Marking | |
-40°C | |
30.48 m x 28.58 mm | |
-40°C to +60°C | |
0.137 mm | |
100 ft. | |
1.125 in. |