Learn More
Stoelting™ Induction Chamber with Sliding Top
Induction Chamber with Sliding Top is for the initial anesthetization of animals difficult to keep masked when awake.
$1242.76 - $1587.97
Specifications
For Use With (Application) Neuroscience
Includes Sliding top, Hook-up kit of 4 ft. of rubber tubing, Inlet port connector
View More Specs
Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity  
10-000-934
View Documents
Stoelting™
53917
9 x 5 x 5 in.
Each for $1,242.76
Only null left
10-000-935
View Documents
Stoelting™
53918
12 x 6 x 6 in.
Each for $1,587.97
Only null left
Description
Description
Route gas through chamber, then to mask or 50206 Scavenger. Includes a hook-up kit of 4 ft. of rubber tubing and inlet port connector.
Specifications
Specifications
Neuroscience
Sliding top, Hook-up kit of 4 ft. of rubber tubing, Inlet port connector
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'