missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-butyric Acid, 97%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor IN12725GM

 View more versions of this product

Catalog No. 18611028


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

133-32-4
Amber Glass Bottle
0.97
1
25 g
Reagent

Chemical Identifiers

133-32-4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.