Learn More
Impacto™ Fingerless Grain Leather Anti-Vibration Gloves with Detachable Wrist Support
Fingerless gloves feature VEP padding in the palm and thumb web area for protection against abrasion, impact, and shock. Detachable wrist support minimizes extension and over-flexion of the wrist to prevent repetitive strain injuries (RSI).
$44.41 - $45.89
Specifications
Cuff Material Grain Leather
Color Blue, Yellow
Material Nylon, Grain Leather
Product Type Fingerless Gloves
Finger Style Fingerless
View More Specs
Products 4
Catalog Number Mfr. No. Size Hand Price Quantity  
Catalog Number Mfr. No. Size Hand Price Quantity  
19-031930
View Documents
Impacto™
70420120021
Small Left
Each for $45.89
Only null left
19-031952
View Documents
Impacto™
70420120022
Small Right
Each for $44.41
Only null left
19-031950
View Documents
Impacto™
70420120032
Medium Right
Each for $44.41
Only null left
19-031948
View Documents
Impacto™
70420120042
Large Right
Each for $44.41
Only null left
Description
Description
  • VEP padding protects the palm and thumb web area from impact
  • Anti-shock wrist support protects from Repetitive Strain Injuries (RSI) such as Carpal Tunnel Syndrome or Tendinitis
  • Wrist support minimizes extension and over flexion of the wrist
  • Provides protection from abrasion, impact, and shock
  • Fingerless style for optimum dexterity
  • Use with hand or electric tools in repetitive tasks
Specifications
Specifications
Grain Leather
Nylon, Grain Leather
Fingerless
704-20
Blue, Yellow
Fingerless Gloves
Yellow
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'