Learn More
Fisherbrand™ Immersion Basket with Handle
Facilitates complete immersion within S-line Ultrasonic Cleaner baths. Fisherbrand™ Immersion Baskets with Handle are manufactured from stainless steel.
Supplier:  Fisherbrand™ 1113390
 View more versions of this product
Catalog No. 29-407-11
Only null left
Explore available promotions
Description
Description
Designed for the complete immersion of items and parts in Fisherbrand™ 112xxx Series Dual Frequency Ultrasonic Cleaners. Choose the Fisherbrand™ Stainless Steel Immersion Basket with Handles designed for the specific ultrasonic cleaner being used.
  • Designed for the Fisherbrand Dual Frequency Ultrasonic Cleaner models
  • Fine mesh bottom keeps small parts secure
  • Two handles for ease-of-use
  • Clean and simple design fits perfectly
  • Top-quality durable stainless steel
Specifications
Specifications
Basket
Ultrasonic Cleaner, 12.75 L
Stainless Steel
Product Suggestions
Product Suggestions